Assay Details
Target Gene Details
Entrez Gene ID: | 12014 |
Gene Name: | BTB and CNC homology 2 |
Gene Aliases: |
E030004N02Rik |
Location: |
Chr.4:32238574-32586108 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bach2 | CJ092609.1 | 1 | 247 |