Assay Details
Target Gene Details
Entrez Gene ID: | 12557 |
Gene Name: | cadherin 17 |
Gene Aliases: |
HPT-1, HPT-1/LI |
Location: |
Chr.4:11758157-11817905 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cdh17 | NM_019753.4 | 15 | 2232 | NP_062727.1 |
AF177669.1 | 15 | 2128 | AAD51125.1 | |
AK089791.1 | 15 | 2200 | ||
AK155633.1 | 15 | 2233 | BAE33356.1 | |
AK171733.1 | 15 | 2155 | BAE42638.1 | |
BC042891.1 | 15 | 2150 | AAH42891.1 | |
D87912.1 | 15 | 2177 | BAB03264.1 |
More Information
Set Membership: |
Intragenic Exonic |