Assay Details
Target Gene Details
Entrez Gene ID: | 70088 |
Gene Name: | MYST/Esa1-associated factor 6 |
Gene Aliases: |
2310005N01Rik, 2810036M01Rik |
Location: |
Chr.4:125085074-125110070 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Meaf6 | NM_001290701.1 | NP_001277630.1 | ||
NM_027310.4 | NP_081586.1 | |||
AK009177.1 | BAB26123.1 | |||
AK012867.1 | ||||
BC108404.1 | AAI08405.1 |