Assay Details
Target Gene Details
Entrez Gene ID: | 230393 |
Gene Name: | focadhesin |
Gene Aliases: |
Kiaa1797, mKIAA1797 |
Location: |
Chr.4:88094630-88411011 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 42 - Exon 43 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Focad | NM_001081184.1 | NP_001074653.1 | ||
AK122556.1 | BAC65838.1 | |||
AK146138.1 | BAE26929.1 | |||
AK146256.1 | BAE27017.1 | |||
BC039622.1 | ||||
BC057079.1 | AAH57079.1 |
Target Gene Details
Entrez Gene ID: | 66775 |
Gene Name: | 3-hydroxyacyl-CoA dehydratase 4 |
Gene Aliases: |
4933428I03Rik, Hcad4, Ptplad2 |
Location: |
Chr.4:88407687-88438935 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hacd4 |