Assay Details
Target Gene Details
Entrez Gene ID: | 67490 |
Gene Name: | UFM1 specific ligase 1 |
Gene Aliases: |
1810074P20Rik, AI429228, Kiaa0776, Maxer, Rcad, mKIAA0776 |
Location: |
Chr.4:25247820-25281926 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ufl1 | NM_026194.4 | 4 | 379 | NP_080470.2 |
AK007993.2 | 4 | 324 | BAB25395.1 | |
AK032663.1 | 4 | 315 | BAC27976.1 | |
AK076206.1 | 4 | 324 | ||
AK129211.1 | 3 | 292 | BAC98021.1 | |
AK132697.1 | 4 | 343 | BAE21308.1 | |
AK207708.1 | ||||
AK208712.1 | ||||
AK218998.1 | ||||
BC132195.1 | 4 | 331 | ||
BC138153.1 | 4 | 331 | ||
BY097965.1 | 4 | 381 |
More Information
Set Membership: |
Intragenic Exonic |