Assay Details
Target Gene Details
Entrez Gene ID: | 230099 |
Gene Name: | carbonic anhydrase 9 |
Gene Aliases: |
CAIX, Ca9, MN/CA9 |
Location: |
Chr.4:43507026-43513810 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Car9 | NM_139305.2 | 8 | 1093 | NP_647466.2 |
AB086322.1 | BAC00816.1 | |||
AJ245857.1 | 8 | 1095 | CAC80975.1 | |
AK136579.1 | 8 | 1094 | BAE23060.1 | |
AK137719.1 | 8 | 1095 | BAE23474.1 | |
AK166403.1 | 5 | 522 | BAE38755.1 | |
BC120544.1 | 8 | 1086 | ||
BC120546.1 | 8 | 1086 | ||
EF122497.1 |