Assay Details
Target Gene Details
Entrez Gene ID: | 78933 |
Gene Name: | ATP/GTP binding protein-like 4 |
Gene Aliases: |
4930578N11Rik, 4931433A01Rik |
Location: |
Chr.4:110397643-111664324 on Build GRCm38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Agbl4 | NM_001048189.4 | NP_001041654.2 | ||
NM_001284190.1 | NP_001271119.1 | |||
NM_030231.3 | NP_084507.1 | |||
AK016502.1 | ||||
DQ867039.1 | ||||
DQ867040.1 | ||||
FN429928.1 |
Target Gene Details
Entrez Gene ID: | 67621 |
Gene Name: | BEN domain containing 5 |
Gene Aliases: |
2310026E23Rik |
Location: |
Chr.4:111414540-111460298 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bend5 | NM_026279.3 | NP_080555.1 | ||
NR_033793.1 | ||||
AK075885.1 | BAC36030.1 | |||
AK156708.1 | BAE33814.1 | |||
AK167425.1 | BAE39513.1 | |||
BC037633.1 | AAH37633.1 | |||
BC051512.1 | AAH51512.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |