Assay Details
Target Gene Details
Entrez Gene ID: | 242341 |
Gene Name: | ATPase, H+ transporting, lysosomal V0 subunit D2 |
Gene Aliases: |
1620401A02Rik, AI324824, V-ATPase |
Location: |
Chr.4:19876838-19922566 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Atp6v0d2 | NM_175406.3 | NP_780615.2 | ||
AB088358.1 | BAC57951.1 | |||
AK075745.1 | BAC35925.1 | |||
AK089507.1 | BAC40907.1 | |||
AK137586.1 | ||||
AK170072.1 | BAE41545.1 | |||
AK170405.1 | BAE41772.1 | |||
AK170578.1 | BAE41890.1 | |||
AK170659.1 | BAE41942.1 | |||
AY145896.1 | AAN61104.1 | |||
AY517482.1 | AAR99405.1 | |||
BC087899.1 | AAH87899.1 |
More Information
Set Membership: |
Intragenic Non-exonic |