Assay Details
Target Gene Details
Entrez Gene ID: | 229228 |
Gene Name: | nudix (nucleoside diphosphate linked moiety X)-type motif 6 |
Gene Aliases: |
Asfgf2b |
Location: |
Chr.3:37404982-37420186 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nudt6 | NM_001291044.1 | 1 | 54 | NP_001277973.1 |
NM_153561.3 | 1 | 54 | NP_705789.1 | |
AF453427.1 | AAN76847.1 | |||
AF453428.1 | AAN76848.1 | |||
AK145299.1 | 1 | 46 | BAE26354.1 | |
BC027267.1 | 1 | 48 | AAH27267.1 | |
BC146000.1 | 1 | 48 |
More Information
Set Membership: |
Intragenic Exonic |