Assay Details
Target Gene Details
Entrez Gene ID: | 67738 |
Gene Name: | peptidylprolyl isomerase D (cyclophilin D) |
Gene Aliases: |
4930564J03Rik, CYP-40, Ppidl, Ppif |
Location: |
Chr.3:79591389-79603650 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ppid | NM_026352.3 | NP_080628.1 | ||
AK003402.1 | BAB22767.1 | |||
AK013919.1 | BAB29056.1 | |||
AK016221.1 | ||||
AK051597.1 | BAC34686.1 | |||
AK142413.1 | ||||
AK150520.1 | BAE29632.1 | |||
AK151091.1 | BAE30104.1 | |||
BC011499.1 | AAH11499.1 | |||
BC019778.1 | AAH19778.1 |
More Information
Set Membership: |
Intragenic Non-exonic |