Assay Details
Target Gene Details
Entrez Gene ID: | 64580 |
Gene Name: | N-deacetylase/N-sulfotransferase (heparin glucosaminyl) 4 |
Gene Aliases: |
4930439H17Rik |
Location: |
Chr.3:125404076-125728351 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ndst4 | NM_022565.2 | NP_072087.2 | ||
AB036838.1 | BAB18517.1 | |||
AK140992.1 |
More Information
Set Membership: |
Intragenic Non-exonic |