Assay Details
Target Gene Details
Entrez Gene ID: | 68911 |
Gene Name: | pygopus 2 |
Gene Aliases: |
1190004M21Rik |
Location: |
Chr.3:89430132-89435131 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pygo2 | NM_001293763.1 | NP_001280692.1 | ||
NM_001293766.1 | NP_001280695.1 | |||
NM_001293767.1 | NP_001280696.1 | |||
NM_001293768.1 | NP_001280697.1 | |||
NM_026869.3 | NP_081145.1 | |||
AK132759.1 | BAE21340.1 | |||
AK202945.1 | ||||
AK205505.1 | ||||
AK208605.1 | ||||
AK209495.1 | ||||
AK212661.1 | ||||
AK216309.1 | ||||
AY486147.1 | AAS55435.1 | |||
BC025640.1 | AAH25640.1 | |||
BQ571639.1 | ||||
BY009144.1 | ||||
BY249701.1 |