Assay Details
Target Gene Details
Entrez Gene ID: | 74838 |
Gene Name: | N(alpha)-acetyltransferase 15, NatA auxiliary subunit |
Gene Aliases: |
5730450D16Rik, 6330400I15, ASTBDN, Narg1, Tbdn-1, mNAT1 |
Location: |
Chr.3:51416016-51475985 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Naa15 | NM_053089.3 | NP_444319.3 | ||
AB221683.1 | ||||
AF510858.1 | AAO33713.1 | |||
BC050017.1 | AAH50017.1 | |||
BQ887090.1 | ||||
CA327064.1 | ||||
CX241349.1 |
More Information
Set Membership: |
Intragenic Non-exonic |