Assay Details
Target Gene Details
Entrez Gene ID: | 59020 |
Gene Name: | PDZ domain containing 1 |
Gene Aliases: |
1700023D20Rik, 2610507N21Rik, 4921513F16Rik, AI267131, AI314638, AL022680, D3Ertd537e, Pdzd1, mPDZK1 |
Location: |
Chr.3:96830065-96870926 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pdzk1 | NM_001146001.1 | 2 | 99 | NP_001139473.1 |
NM_021517.2 | 2 | 145 | NP_067492.2 | |
AF220100.1 | 2 | 84 | AAF73863.1 | |
AF474247.1 | 1 | 83 | AAL84634.1 | |
AK006269.1 | 2 | 619 | BAB24493.1 | |
AK012070.1 | 2 | 135 | BAB28007.1 | |
AK014879.1 | 2 | 402 | BAB29600.1 | |
AK029752.1 | 2 | 603 | BAC26598.1 | |
AK029764.1 | 2 | 613 | BAC26605.1 | |
AK165415.1 | 2 | 80 | ||
AW259368.1 | 2 | 145 | ||
BC013512.1 | 1 | 68 | AAH13512.1 |