Assay Details
Target Gene Details
Entrez Gene ID: | 67583 |
Gene Name: | RIKEN cDNA 4930442L01 gene |
Gene Aliases: |
- |
Location: |
Chr.3:96829826-96832070 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
4930442L01Rik | NR_015596.1 | 2 | 1812 | |
AK006795.1 | 1 | 1987 | ||
AK015364.1 | 2 | 1812 |
Target Gene Details
Entrez Gene ID: | 59020 |
Gene Name: | PDZ domain containing 1 |
Gene Aliases: |
1700023D20Rik, 2610507N21Rik, 4921513F16Rik, AI267131, AI314638, AL022680, D3Ertd537e, Pdzd1, mPDZK1 |
Location: |
Chr.3:96830065-96870926 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pdzk1 | NM_021517.2 | NP_067492.2 | ||
AK012070.1 | BAB28007.1 | |||
AW259368.1 |