Assay Details
Target Gene Details
Entrez Gene ID: | 329650 |
Gene Name: | mediator complex subunit 12-like |
Gene Aliases: |
A130035F20, A630079M02, AU044581, TRALPUSH |
Location: |
Chr.3:59005595-59318416 on Build GRCm38 |
Assay Gene Location: | Within Intron 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Med12l | NM_177855.3 | NP_808523.2 | ||
AK047743.1 | BAC33144.1 | |||
BP757618.1 |
Target Gene Details
Entrez Gene ID: | 140795 |
Gene Name: | purinergic receptor P2Y, G-protein coupled, 14 |
Gene Aliases: |
A330108O13Rik, Gpr105, P2Y14 |
Location: |
Chr.3:59114623-59153492 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
P2ry14 | NM_001008497.2 | NP_001008497.1 | ||
NM_001287119.1 | NP_001274048.1 | |||
NM_001287120.1 | NP_001274049.1 | |||
NM_001287121.1 | NP_001274050.1 | |||
NM_001287122.1 | NP_001274051.1 | |||
NM_001287123.1 | NP_001274052.1 | |||
NM_001287124.1 | NP_001274053.1 | |||
NM_133200.4 | NP_573463.1 | |||
AF177211.1 | AAG09275.1 | |||
AK039791.1 | BAC30456.1 | |||
AK140301.1 | BAE24324.1 | |||
AK155035.1 | ||||
AK164982.1 | BAE37990.1 | |||
BC028995.1 | AAH28995.1 | |||
BC058558.1 | AAH58558.1 | |||
BY177387.1 | ||||
CF169635.1 | ||||
CK622172.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |