Assay Details
Target Gene Details
Entrez Gene ID: | 18857 |
Gene Name: | peripheral myelin protein 2 |
Gene Aliases: |
P2 |
Location: |
Chr.3:10179851-10183885 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pmp2 | NM_001030305.2 | NP_001025476.1 | ||
BC099520.1 | AAH99520.1 |