Assay Details
Target Gene Details
Entrez Gene ID: | 54635 |
Gene Name: | platelet-derived growth factor, C polypeptide |
Gene Aliases: |
1110064L01Rik, AI647969, PDGF-C |
Location: |
Chr.3:81035885-81214040 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pdgfc | NM_019971.2 | NP_064355.1 | ||
AF117608.1 | AAF22516.1 | |||
AF266467.1 | AAK58566.1 | |||
AF286725.1 | AAF91483.1 | |||
AK033734.1 | BAC28455.1 | |||
AK042767.1 | BAC31358.1 | |||
AK052947.1 | BAC35216.1 | |||
AK081347.1 | ||||
BC037696.1 | AAH37696.1 |
More Information
Set Membership: |
Intragenic Non-exonic |