Assay Details
Target Gene Details
Entrez Gene ID: | 27371 |
Gene Name: | SH2 domain containing 2A |
Gene Aliases: |
Lad, Ribp, Tsad |
Location: |
Chr.3:87846755-87855722 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sh2d2a | NM_001025571.2 | 5 | 866 | NP_001020742.1 |
NM_021309.3 | 5 | 866 | NP_067284.1 | |
AF203343.1 | 5 | 873 | AAF19396.1 | |
AK037489.1 | 5 | 588 | BAC29819.1 | |
AK172182.1 | 5 | 618 | BAE42870.1 | |
BC034847.1 | 5 | 598 | AAH34847.1 | |
U69490.1 | 5 | 576 | AAB58422.1 |
More Information
Set Membership: |
Intragenic Exonic |