Assay Details
Target Gene Details
Entrez Gene ID: | 242202 |
Gene Name: | phosphodiesterase 5A, cGMP-specific |
Gene Aliases: |
Cgbpde, Cn5n, Pde5, Pde5a1 |
Location: |
Chr.3:122729158-122859374 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pde5a | NM_153422.2 | 8 | 1316 | NP_700471.2 |
AF541937.1 | 8 | 1316 | AAN17330.1 | |
BC119137.1 | 8 | 1279 | ||
BC120486.1 | 8 | 1279 |
More Information
Set Membership: |
Intragenic Exonic |