Assay Details
Target Gene Details
Entrez Gene ID: | 108943 |
Gene Name: | tRNA methyltransferase 10A |
Gene Aliases: |
3110023L08Rik, AA794508, Rg9mtd2, Rnmtd2 |
Location: |
Chr.3:138143517-138159821 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Trmt10a | AK089967.1 | 1 | 45 | BAC41020.1 |
AK145681.1 | ||||
AK146481.1 | ||||
BC098478.1 | AAH98478.1 |
More Information
Set Membership: |
Intragenic Non-exonic |