Assay Details
Target Gene Details
Entrez Gene ID: | 73251 |
Gene Name: | SET domain containing (lysine methyltransferase) 7 |
Gene Aliases: |
1600028F23Rik, H3K4MT, KMT7, Set7, Set7/9, mKIAA1717 |
Location: |
Chr.3:51515318-51560823 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Setd7 | NM_080793.5 | 3 | 538 | NP_542983.3 |
AF448509.1 | 3 | 320 | AAL56578.1 | |
AK048924.1 | 3 | 375 | BAC33493.1 | |
AK129428.1 | 3 | 327 | BAC98238.1 | |
AK147316.1 | 3 | 594 | ||
AK147413.1 | 3 | 364 | BAE27897.1 | |
AK147422.1 | 3 | 539 | BAE27903.1 | |
AK147667.1 | 3 | 573 | BAE28059.1 | |
AK170161.1 | 3 | 382 | BAE41607.1 | |
BC050190.1 | 3 | 362 | AAH50190.1 |
More Information
Set Membership: |
Intragenic Exonic |