Assay Details
Target Gene Details
Entrez Gene ID: | 19142 |
Gene Name: | protease, serine 12 neurotrypsin (motopsin) |
Gene Aliases: |
Bssp-3, motopsin |
Location: |
Chr.3:123446913-123506602 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Prss12 | NM_008939.2 | NP_032965.1 | ||
AK132633.1 | ||||
AK206246.1 | ||||
AK208903.1 | ||||
BC031429.1 | AAH31429.1 | |||
D89871.1 | BAA23986.1 | |||
Y13192.1 | CAA73646.1 |