Assay Details
Target Gene Details
Entrez Gene ID: | 67547 |
Gene Name: | solute carrier family 39 (metal ion transporter), member 8 |
Gene Aliases: |
4933419D20Rik, AA986696, BIGM103, ZIP-8, Zip8 |
Location: |
Chr.3:135816883-135888572 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc39a8 | NM_001135149.1 | 6 | 1097 | NP_001128621.1 |
NM_001135150.1 | 6 | 1247 | NP_001128622.1 | |
NM_026228.5 | 6 | 1206 | NP_080504.3 | |
AK014895.1 | 6 | 1210 | BAB29610.1 | |
AK016853.1 | 6 | 984 | BAB30465.1 | |
AK028215.1 | 6 | 995 | ||
AK085740.1 | 6 | 1101 | BAC39526.1 | |
AK089060.1 | 5 | 1106 | BAC40727.1 | |
BC006731.1 | 5 | 908 | AAH06731.1 |
More Information
Set Membership: |
Intragenic Exonic |