Assay Details
Target Gene Details
Entrez Gene ID: | 66350 |
Gene Name: | phospholipase A2, group XIIA |
Gene Aliases: |
2310004B05Rik, GXII, GXII-1, GXII-1-PLA2, Pla2g12, Rossy, mGXII-1 |
Location: |
Chr.3:129878606-129895825 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pla2g12a | NM_001286948.1 | 3 | 455 | NP_001273877.1 |
NM_023196.4 | 3 | 455 | NP_075685.2 | |
NM_183423.3 | 3 | 238 | NP_904359.1 | |
NR_104611.1 | 3 | 455 | ||
AK003183.1 | 3 | 415 | ||
AK009133.1 | 3 | 457 | BAB26094.1 | |
AK010011.1 | 3 | 445 | BAB26641.1 | |
AK010174.1 | 3 | 421 | BAB26747.1 | |
AK012414.1 | 3 | 383 | BAC25368.1 | |
AK163693.1 | 3 | 369 | BAE37460.1 | |
AK178670.1 | ||||
AY007381.1 | 3 | 428 | AAG23336.1 | |
AY007382.1 | 3 | 238 | AAG23337.1 | |
BC026812.1 | 3 | 420 | AAH26812.1 | |
BC051117.1 | 3 | 455 | AAH51117.1 |
More Information
Set Membership: |
Intragenic Exonic |