Assay Details
Target Gene Details
Entrez Gene ID: | 53814 |
Gene Name: | ornithine decarboxylase antizyme 3 |
Gene Aliases: |
AZ3, Az-3, Oaz-t |
Location: |
Chr.3:94433387-94436651 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Oaz3 | NM_016901.3 | 3 | 566 | NP_058597.2 |
AB045835.1 | 3 | 491 | BAB97207.1 | |
AF175297.1 | 3 | 531 | AAD51735.1 | |
AI449747.1 | ||||
AK006116.1 | 3 | 501 | ||
AK006634.1 | 3 | 502 | ||
AK076942.1 | 3 | 508 |
More Information
Set Membership: |
Intragenic Exonic |