Assay Details
Target Gene Details
Entrez Gene ID: | 78523 |
Gene Name: | mitochondrial ribosomal protein L9 |
Gene Aliases: |
8030480E20Rik, AA409733, C330013D18Rik, L9mt |
Location: |
Chr.3:94443272-94448708 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mrpl9 | NM_030116.2 | NP_084392.1 | ||
AB049637.1 | BAB40842.1 | |||
AK021196.1 | ||||
AK040055.1 | BAC30503.1 | |||
AK046099.1 | BAC32604.1 | |||
AK161563.1 | ||||
AK162023.1 | BAE36685.1 | |||
BC116385.1 | ||||
BC116386.1 |
More Information
Set Membership: |
Intragenic Non-exonic |