Assay Details
Target Gene Details
Entrez Gene ID: | 14862 |
Gene Name: | glutathione S-transferase, mu 1 |
Gene Aliases: |
Gstb-1, Gstb1 |
Location: |
Chr.3:108012250-108017973 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gstm1 | NM_010358.5 | NP_034488.1 | ||
BC003822.1 | AAH03822.1 | |||
BC046758.1 | AAH46758.1 | |||
BC091763.1 | AAH91763.1 | |||
J03952.1 | AAA37747.1 | |||
J04632.1 | AAA37705.1 |
More Information
Set Membership: |
Intragenic Non-exonic |