Assay Details
Target Gene Details
Entrez Gene ID: | 208691 |
Gene Name: | eukaryotic translation initiation factor 5A2 |
Gene Aliases: |
9630038B20, eIF5AII |
Location: |
Chr.3:28781311-28798846 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eif5a2 | NM_177586.5 | NP_808254.1 | ||
AK036127.1 | ||||
AK052412.1 | BAC34978.1 | |||
AK082214.1 | BAC38441.1 | |||
AY205262.1 | AAO18680.1 | |||
AY205263.1 | AAO18681.1 | |||
AY205264.1 | AAO18682.1 | |||
AY205265.1 | AAO18683.1 | |||
BC106843.2 | AAI06844.1 | |||
BC106844.1 | AAI06845.1 |
More Information
Set Membership: |
Intragenic Non-exonic |