Assay Details
Target Gene Details
Entrez Gene ID: | 13645 |
Gene Name: | epidermal growth factor |
Gene Aliases: |
AI790464 |
Location: |
Chr.3:129677569-129755322 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Egf | NM_010113.3 | 4 | 964 | NP_034243.2 |
AK084927.1 | 4 | 959 | ||
AK085707.1 | 4 | 531 | ||
AK142666.1 | 4 | 660 | ||
BC017681.1 | 5 | 901 | AAH17681.1 | |
BC060741.1 | 4 | 949 | AAH60741.1 | |
BC092277.1 | 4 | 949 | AAH92277.1 | |
J00380.1 | 4 | 959 | AAA37539.1 | |
V00741.1 | 4 | 915 | CAA24115.1 |
More Information
Set Membership: |
Intragenic Exonic |