Assay Details
Target Gene Details
Entrez Gene ID: | 20585 |
Gene Name: | helicase-like transcription factor |
Gene Aliases: |
AF010600, BC057116, P113, Smarca3, Snf2l3 |
Location: |
Chr.3:20057811-20118491 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hltf | NM_009210.3 | 1 | 60 | NP_033236.2 |
NR_105046.1 | 1 | 60 | ||
AF010600.1 | 1 | 60 | AAB63915.1 | |
AF165911.1 | ||||
AK141235.1 | BAE24607.1 | |||
AK142961.1 | BAE25232.1 | |||
AK166911.1 | ||||
BC057116.1 | AAH57116.1 | |||
BC059240.1 | AAH59240.1 | |||
BY007037.1 | 1 | 67 | ||
BY205383.1 | 1 | 61 | ||
BY726838.1 |