Assay Details
Target Gene Details
Entrez Gene ID: | 53975 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 20 |
Gene Aliases: |
GEMIN3, dp103 |
Location: |
Chr.3:105678270-105688113 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddx20 | NM_017397.3 | NP_059093.3 | ||
AF217000.1 | AAK56847.1 | |||
AF217001.1 | AAK56848.1 | |||
AF220454.1 | AAF76301.1 | |||
AJ250027.1 | CAB86201.1 | |||
AK154518.1 | ||||
BC125471.1 | ||||
BC137721.1 |