Assay Details
Target Gene Details
Entrez Gene ID: | 14468 |
Gene Name: | guanylate binding protein 2b |
Gene Aliases: |
Gbp-1, Gbp1, Mag-1, Mpa-1, Mpa1 |
Location: |
Chr.3:142594816-142619179 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gbp2b | NM_010259.2 | 10 | 1685 | NP_034389.2 |
AK077796.1 | 10 | 1687 | ||
EF494422.1 | 10 | 1685 |
More Information
Set Membership: |
Intragenic Exonic |