Assay Details
Target Gene Details
Entrez Gene ID: | 12349 |
Gene Name: | carbonic anhydrase 2 |
Gene Aliases: |
AI131712, CAII, Ca2, Car-2, Ltw-5, Lvtw-5 |
Location: |
Chr.3:14886278-14900770 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Car2 | NM_009801.4 | NP_033931.4 | ||
AK002333.1 | BAB22019.1 | |||
AK002498.1 | BAB22146.2 | |||
AK146015.1 | ||||
BC055291.1 | AAH55291.1 | |||
BU946040.1 | ||||
K00811.1 | AAA37356.1 |