Assay Details
Target Gene Details
Entrez Gene ID: | 11770 |
Gene Name: | fatty acid binding protein 4, adipocyte |
Gene Aliases: |
422/aP2, ALBP/Ap2, Ap2, Lbpl |
Location: |
Chr.3:10204342-10208597 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fabp4 | NM_024406.2 | 4 | 489 | NP_077717.1 |
AA066537.1 | 1 | 158 | ||
AK003143.1 | 4 | 490 | BAB22601.1 | |
AK088535.1 | 4 | 490 | BAC40410.1 | |
BC002148.1 | 1 | 1904 | AAH02148.1 | |
BC054426.1 | 4 | 472 | AAH54426.1 | |
CT010390.1 | CAJ18597.1 | |||
K02109.1 | 4 | 467 | AAA39416.1 | |
M28726.1 | 4 | 394 | AAA37112.1 |