Assay Details
Target Gene Details
Entrez Gene ID: | 99662 |
Gene Name: | EPS8-like 3 |
Gene Aliases: |
AI504489, Eps8r3 |
Location: |
Chr.3:107873860-107892900 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eps8l3 | NM_133867.2 | NP_598628.1 | ||
AK136584.1 | ||||
AK167630.1 | BAE39681.1 | |||
AY074932.1 | AAL76121.1 | |||
BC014734.1 | AAH14734.1 |
More Information
Set Membership: |
Intragenic Non-exonic |