Assay Details
Target Gene Details
Entrez Gene ID: | 329628 |
Gene Name: | FAT tumor suppressor homolog 4 (Drosophila) |
Gene Aliases: |
6030410K14Rik, 9430004M15 |
Location: |
Chr.3:38886487-39011987 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fat4 | NM_183221.3 | 14 | 12560 | NP_899044.3 |
BC079887.1 | 6 | 3654 | AAH79887.1 | |
DQ286572.1 | 14 | 12560 | ABB88946.1 |
More Information
Set Membership: |
Intragenic Exonic |