Assay Details
Target Gene Details
Entrez Gene ID: | 21818 |
Gene Name: | transglutaminase 3, E polypeptide |
Gene Aliases: |
AI893889, TG E, TG(E), TGE, TGase E, TGase-3 |
Location: |
Chr.2:129987794-130050399 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tgm3 | NM_009374.3 | 3 | 365 | NP_033400.2 |
BC129890.1 | 3 | 331 | ||
BC129891.1 | 3 | 331 | ||
L10385.1 | 3 | 356 | AAA40421.1 |
More Information
Set Membership: |
Intragenic Exonic |