Assay Details
Target Gene Details
Entrez Gene ID: | 21817 |
Gene Name: | transglutaminase 2, C polypeptide |
Gene Aliases: |
G[a]h, TG2, TGase2, tTG, tTGas |
Location: |
Chr.2:158116405-158146392 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tgm2 | NM_009373.3 | 3 | 323 | NP_033399.1 |
AF076928.1 | 2 | 238 | AAC62014.1 | |
AF114266.1 | 3 | 288 | AAD37501.1 | |
AK052912.1 | 3 | 298 | BAC35200.1 | |
AK054108.1 | 3 | 298 | ||
AK080224.1 | 3 | 323 | BAC37852.1 | |
AK080593.1 | 3 | 298 | BAC37952.1 | |
AK086001.1 | 3 | 298 | ||
AK089481.1 | 3 | 326 | BAC40899.1 | |
AK143712.1 | 3 | 359 | BAE25511.1 | |
AK151776.1 | 3 | 324 | BAE30682.1 | |
AK152152.1 | 3 | 298 | BAE30987.1 | |
AK152627.1 | 3 | 324 | BAE31370.1 | |
AK159255.1 | 3 | 298 | BAE34936.1 | |
AK166302.1 | 3 | 298 | BAE38690.1 | |
AK168990.1 | 3 | 299 | BAE40790.1 | |
AK169356.1 | 3 | 326 | BAE41105.1 | |
BC016492.1 | 3 | 264 | AAH16492.1 | |
M55154.1 | 3 | 260 | AAA40420.1 |
More Information
Set Membership: |
Intragenic Exonic |