Assay Details
Target Gene Details
Entrez Gene ID: | 21817 |
Gene Name: | transglutaminase 2, C polypeptide |
Gene Aliases: |
G[a]h, TG2, TGase2, tTG, tTGas |
Location: |
Chr.2:158116405-158146392 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tgm2 | NM_009373.3 | NP_033399.1 | ||
AF076928.1 | AAC62014.1 | |||
AF114266.1 | AAD37501.1 | |||
AK052912.1 | BAC35200.1 | |||
AK080224.1 | BAC37852.1 | |||
AK080593.1 | BAC37952.1 | |||
AK086001.1 | ||||
AK089481.1 | BAC40899.1 | |||
AK143712.1 | BAE25511.1 | |||
AK151776.1 | BAE30682.1 | |||
AK152152.1 | BAE30987.1 | |||
AK152627.1 | BAE31370.1 | |||
AK159255.1 | BAE34936.1 | |||
AK166302.1 | BAE38690.1 | |||
AK168990.1 | BAE40790.1 | |||
AK169356.1 | BAE41105.1 | |||
BC016492.1 | AAH16492.1 | |||
M55154.1 | AAA40420.1 |
More Information
Set Membership: |
Intragenic Non-exonic |