Assay Details
Target Gene Details
Entrez Gene ID: | 228012 |
Gene Name: | tousled-like kinase 1 |
Gene Aliases: |
4930545J15Rik |
Location: |
Chr.2:70712407-70825480 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tlk1 | NM_172664.3 | NP_766252.2 | ||
AK029773.1 | BAC26610.1 | |||
AK172897.1 | BAD32175.1 | |||
BC033931.1 | ||||
BC051641.1 | AAH51641.1 |
More Information
Set Membership: |
Intragenic Non-exonic |