Assay Details
Target Gene Details
Entrez Gene ID: | 19668 |
Gene Name: | recombination signal binding protein for immunoglobulin kappa J region-like |
Gene Aliases: |
RBP-L, Rbpsuhl |
Location: |
Chr.2:164403056-164415448 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rbpjl | NM_009036.1 | NP_033062.1 | ||
AK052449.1 | 3 | 1374 | ||
AK131814.1 | BAE20815.1 | |||
AK131832.1 | ||||
BC152311.1 | ||||
Y10926.1 | CAA71856.1 |
More Information
Set Membership: |
Intragenic Non-exonic |