Assay Details
Target Gene Details
Entrez Gene ID: | 269397 |
Gene Name: | synovial sarcoma translocation gene on chromosome 18-like 1 |
Gene Aliases: |
A230053O16Rik, CREST |
Location: |
Chr.2:180042483-180070201 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ss18l1 | NM_178750.5 | NP_848865.4 | ||
AK038669.1 | BAC30089.1 | |||
AK054560.1 | BAC35824.1 | |||
AK133944.1 | BAE21942.1 | |||
AK173004.1 | BAD32282.1 | |||
BC053087.1 | AAH53087.1 | |||
BC054730.1 | AAH54730.1 |
More Information
Set Membership: |
Intragenic Non-exonic |