Assay Details
Target Gene Details
Entrez Gene ID: | 27054 |
Gene Name: | SEC23B (S. cerevisiae) |
Gene Aliases: |
- |
Location: |
Chr.2:144556229-144590753 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sec23b | NM_001252543.1 | 2 | 264 | NP_001239472.1 |
NM_001252544.1 | 2 | 257 | NP_001239473.1 | |
NM_001252545.1 | 2 | 246 | NP_001239474.1 | |
NM_019787.4 | 2 | 275 | NP_062761.2 | |
AF200326.1 | 1 | 147 | AAF08301.1 | |
AK014597.1 | 2 | 225 | BAB29452.1 | |
AK139729.1 | 2 | 189 | ||
AK146083.1 | 2 | 224 | BAE26887.1 | |
AK149693.1 | 2 | 204 | BAE29030.1 | |
AK153214.1 | 2 | 231 | BAE31812.1 | |
AK155168.1 | 2 | 205 | BAE33090.1 | |
AK158398.1 | 2 | 242 | BAE34488.1 | |
AK166330.1 | 2 | 207 | BAE38710.1 | |
AK167382.1 | 2 | 213 | ||
AK167732.1 | 2 | 193 | BAE39773.1 | |
AK171597.1 | 2 | 205 | BAE42552.1 | |
AK179166.1 | 1 | 66 | ||
BC005464.1 | 2 | 200 | AAH05464.1 | |
BC011160.1 | 2 | 227 | AAH11160.1 | |
BY110125.1 | 2 | 259 | ||
BY127982.1 | 2 | 232 |
More Information
Set Membership: |
Intragenic Exonic |