Assay Details
Target Gene Details
Entrez Gene ID: | 78239 |
Gene Name: | serine protease inhibitor, Kunitz type 4 |
Gene Aliases: |
9230105I15Rik |
Location: |
Chr.2:164698501-164702448 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Spint4 | NM_030058.2 | 4 | 604 | NP_084334.1 |
AK020315.1 | BAB32069.1 | |||
AY226988.1 | 4 | 604 | AAP43945.1 | |
BC119299.1 | ||||
BC119301.1 |
More Information
Set Membership: |
Intragenic Exonic |