Assay Details
Target Gene Details
Entrez Gene ID: | 75526 |
Gene Name: | epididymal peptidase inhibitor |
Gene Aliases: |
1700024E17Rik, Spinlw1 |
Location: |
Chr.2:164588343-164593571 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eppin | NM_029325.2 | NP_083601.1 | ||
AF346413.2 | AAK31335.1 | |||
AK006296.1 | BAB24514.1 | |||
BC048637.1 | AAH48637.1 |