Assay Details
Target Gene Details
Entrez Gene ID: | 241308 |
Gene Name: | Ral GEF with PH domain and SH3 binding motif 1 |
Gene Aliases: |
5830418G11Rik, AI853783, AI854138, RALGEF2, RALGPS1A, RalGEF 2, mKIAA0351 |
Location: |
Chr.2:33133419-33371511 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ralgps1 | NM_001290570.1 | NP_001277499.1 | ||
NM_001290572.1 | NP_001277501.1 | |||
NM_175211.5 | NP_780420.1 | |||
AK017939.1 | BAC25535.1 | |||
AK076767.1 | BAC36473.1 | |||
AK129123.1 | BAC97933.1 | |||
AK170248.1 | ||||
BC072656.1 | AAH72656.1 |
More Information
Set Membership: |
Intragenic Non-exonic |