Assay Details
Target Gene Details
Entrez Gene ID: | 77799 |
Gene Name: | Src-like-adaptor 2 |
Gene Aliases: |
A930009E21Rik, AI430952, SLAP-2, SLAP2 |
Location: |
Chr.2:156872922-156887241 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sla2 | NM_029983.5 | NP_084259.1 | ||
AF287467.1 | AAL38196.1 | |||
AF434990.1 | AAL86403.1 | |||
AK020837.1 | BAB32223.1 | |||
AK030877.1 | BAC27168.1 | |||
AK088672.1 | BAC40495.1 | |||
AK138709.1 | BAE23754.1 | |||
BC052655.1 | AAH52655.1 |
More Information
Set Membership: |
Intragenic Non-exonic |