Assay Details
Target Gene Details
Entrez Gene ID: | 19281 |
Gene Name: | protein tyrosine phosphatase, receptor type, T |
Gene Aliases: |
R-PTP-T, RPTP-rfo, RPTP-rho, RPTPmam4, RPTPrho, mKIAA0283, mRPTPrho |
Location: |
Chr.2:161521988-162661147 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptprt | NM_001291149.1 | 10 | 1966 | NP_001278078.1 |
NM_001291150.1 | 10 | 1966 | NP_001278079.1 | |
NM_001291151.1 | 10 | 1966 | NP_001278080.1 | |
NM_021464.5 | 10 | 1966 | NP_067439.1 | |
AF152556.4 | 10 | 1887 | AAD34158.4 | |
AF162856.2 | 10 | 1889 | AAF82400.2 | |
AF162857.1 | 10 | 1889 | AAF82401.1 | |
AF244125.1 | 11 | 1967 | AAF44712.1 | |
AK220254.1 | 3 | 519 | BAD90179.1 | |
AY026861.1 | 11 | 1718 | AAK18741.1 | |
AY026862.1 | 11 | 1718 | AAK18742.1 | |
AY026863.1 | 11 | 1718 | AAK18743.1 |
More Information
Set Membership: |
Intragenic Exonic |