Assay Details
Target Gene Details
Entrez Gene ID: | 94249 |
Gene Name: | solute carrier family 24 (sodium/potassium/calcium exchanger), member 3 |
Gene Aliases: |
NCKX3 |
Location: |
Chr.2:145167712-145642166 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc24a3 | NM_053195.2 | 13 | 1316 | NP_444425.1 |
AF314821.1 | 13 | 1356 | AAG60049.1 | |
BC005742.1 | 3 | 446 | AAH05742.1 | |
BC017615.1 | 13 | 1316 | AAH17615.1 |
More Information
Set Membership: |
Intragenic Exonic |